Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522408_m1
          See other CYP2U1 GE Assays ›
          Gene Symbol:
          Cyp2u1
          Gene Name:
          cytochrome P450, family 2, subfamily u, polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 236414131 - 236431686 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522408_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2u1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024779.1 NP_001019950.1 1 - 2 519 68
          XM_006233310.2 XP_006233372.1 1 - 2 607 68
          XM_008761499.1 XP_008759721.1 1 - 2 609 68
          XM_008761500.1 XP_008759722.1 1 - 2 607 68
          GenBank mRNA BC097442.1 - 1 - 2 519 7456028 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004704.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2u1

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          organic acid metabolic process
          xenobiotic metabolic process
          exogenous drug catabolic process
          oxidation-reduction process
          iron ion binding
          steroid hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P119264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P119264 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P119264 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P119264 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P119264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96797 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20470 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826844 - -
          Affymetrix Rat Gene 1.0 ST Array 10826845 - -
          Affymetrix Rat Gene 1.0 ST Array 10826843 - -
          Affymetrix Rat Gene 1.0 ST Array 10826838 - -
          Affymetrix Rat Gene 1.0 ST Array 10826840 - -
          Affymetrix Rat Gene 1.0 ST Array 10826842 - -
          Affymetrix Rat Gene 1.0 ST Array 10826841 - -
          Affymetrix Rat Gene 1.0 ST Array 10826839 - -
          Affymetrix Rat Gene 1.1 ST Array 10826837 - -
          Rat Genome 230 2.0 Array 1398710_PM_at - -
          Rat Genome 230 2.0 Array 1398710_at - -
          Affymetrix HT Rat Focus Array 1398710_at - -
          Affymetrix HT Rat Focus Array 1398710_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200032734.rn.1 TC0200004704.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032735.rn.1 TC0200004704.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032732.rn.1 TC0200004704.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032733.rn.1 TC0200004704.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032736.rn.1 TC0200004704.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200032728.rn.1 TC0200004704.rn.1 RTA TC
          Rat Expression Set 230 1398710_PM_at - -
          Rat Expression Set 230 1398710_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826837 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20917308 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373501 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline