Search
Search
| Gene Symbol: Ranbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244357.2 | XP_006244419.1 | 10 - 11 | 968 | 77 | |
| XM_006244358.2 | XP_006244420.1 | 9 - 10 | 940 | 77 | ||
| XM_006244359.2 | XP_006244421.1 | 7 - 8 | 782 | 77 | ||
| XM_006244360.2 | XP_006244422.1 | 9 - 10 | 763 | 77 | ||
| XM_006244361.2 | XP_006244423.1 | 10 - 11 | 971 | 77 | ||
| XM_006244362.2 | XP_006244424.1 | 6 - 7 | 577 | 77 | ||
| XM_576696.6 | XP_576696.4 | 8 - 9 | 735 | 77 | ||
Set Membership: |
|