Search
Search

Tmem198b| Gene Symbol: Tmem198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109281.1 | NP_001102751.1 | 3 - 4 | 1241 | 65 | |
| XM_006240794.2 | XP_006240856.1 | 3 - 4 | 1102 | 65 | ||
| XM_006240795.1 | XP_006240857.1 | 2 - 3 | 1156 | 65 | ||
| XM_008765038.1 | XP_008763260.1 | 2 - 3 | 1546 | 65 | ||
Set Membership: |
|