Search
Search
| Gene Symbol: Adm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201426.1 | NP_958829.1 | 2 - 3 | 278 | 57 | |
| GenBank mRNA | AB121036.1 | - | 2 - 3 | 278 | 57 | |
| AB181297.1 | - | 2 - 3 | 287 | 57 | ||
| AY590103.1 | - | 1 - 2 | 106 | 57 | ||
| FQ234235.1 | - | 2 - 3 | 271 | 57 | ||
Set Membership: |
|