Search
Search
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012910.2 | NP_037042.1 | 5 - 6 | 382 | 65 | |
| XM_006229734.2 | XP_006229796.1 | 5 - 6 | 1008 | 65 | ||
| XM_006229735.2 | XP_006229797.1 | 5 - 6 | 1007 | 65 | ||
| XM_006229736.2 | XP_006229798.1 | 5 - 6 | 977 | 65 | ||
| XM_006229737.2 | XP_006229799.1 | 5 - 6 | 976 | 65 | ||
| XM_006229738.2 | XP_006229800.1 | 5 - 6 | 1009 | 65 | ||
| XM_006229739.2 | XP_006229801.1 | 5 - 6 | 1124 | 65 | ||
| XM_006229740.2 | XP_006229802.1 | 5 - 6 | 510 | 65 | ||
| XM_006229741.2 | XP_006229803.1 | 5 - 6 | 523 | 65 | ||
| GenBank mRNA | M91589.1 | - | 5 - 6 | 385 | 65 | |
Set Membership: |
|