Search
Search
| Gene Symbol: Tecpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240617.2 | XP_006240679.1 | 17 - 18 | 4280 | 64 | |
| XM_006240618.2 | XP_006240680.1 | 17 - 18 | 3984 | 64 | ||
| XM_006240620.2 | XP_006240682.1 | 16 - 17 | 3696 | 64 | ||
| XM_008764970.1 | XP_008763192.1 | 18 - 19 | 4047 | 64 | ||
| XM_008764971.1 | XP_008763193.1 | 15 - 16 | 3723 | 64 | ||
| XM_008764972.1 | XP_008763194.1 | 16 - 17 | 3666 | 64 | ||
Set Membership: |
|