Search
Search
| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107070.1 | NP_001100540.1 | 7 - 8 | 1171 | 64 | |
| XM_006247803.2 | XP_006247865.1 | 7 - 8 | 1158 | 64 | ||
| XM_006247804.2 | XP_006247866.1 | 7 - 8 | 983 | 64 | ||
Set Membership: |
|