Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01754023_m1
          See other NXF7 GE Assays ›
          Gene Symbol:
          Nxf7
          Gene Name:
          nuclear RNA export factor 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 106049075 - 106066342 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01754023_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nxf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037216.1 NP_001032293.1 20 - 21 1931 110
          XM_006257254.2 XP_006257316.1 20 - 21 1964 110
          XM_008773431.1 XP_008771653.1 20 - 21 2502 110
          GenBank mRNA BC100151.1 - 20 - 21 1931 7456178 110
          FQ229456.1 - 9 - 10 758 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002199.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nxf7

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA export from nucleus
          nucleotide binding
          mRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56862 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17810 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17810 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935018 - -
          Affymetrix Rat Gene 1.0 ST Array 10935007 - -
          Affymetrix Rat Gene 1.0 ST Array 10935011 - -
          Affymetrix Rat Gene 1.0 ST Array 10935009 - -
          Affymetrix Rat Gene 1.0 ST Array 10935013 - -
          Affymetrix Rat Gene 1.0 ST Array 10935020 - -
          Affymetrix Rat Gene 1.0 ST Array 10935003 - -
          Affymetrix Rat Gene 1.0 ST Array 10935014 - -
          Affymetrix Rat Gene 1.0 ST Array 10935006 - -
          Affymetrix Rat Gene 1.0 ST Array 10935002 - -
          Affymetrix Rat Gene 1.0 ST Array 10935017 - -
          Affymetrix Rat Gene 1.0 ST Array 10935016 - -
          Affymetrix Rat Gene 1.0 ST Array 10935000 - -
          Affymetrix Rat Gene 1.0 ST Array 10935010 - -
          Affymetrix Rat Gene 1.0 ST Array 10935008 - -
          Affymetrix Rat Gene 1.0 ST Array 10935004 - -
          Affymetrix Rat Gene 1.0 ST Array 10935005 - -
          Affymetrix Rat Gene 1.0 ST Array 10935019 - -
          Affymetrix Rat Gene 1.0 ST Array 10934999 - -
          Affymetrix Rat Gene 1.0 ST Array 10935015 - -
          Affymetrix Rat Gene 1.0 ST Array 10935012 - -
          Affymetrix Rat Gene 1.1 ST Array 10934998 - -
          Rat Genome 230 2.0 Array 1392780_at - -
          Rat Genome 230 2.0 Array 1392780_PM_at - -
          Affymetrix HT Rat Focus Array 1392780_PM_at - -
          Affymetrix HT Rat Focus Array 1392780_at - -
          Affymetrix Rat Clariom S Assay PSR0X00013800.rn.1 TC0X00002199.rn.1 RTA TC
          Rat Expression Set 230 1392780_PM_at - -
          Rat Expression Set 230 1392780_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934998 - -
          Rat Genome U34 Set rc_AI233520_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22364959 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367173 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362907 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline