Search
Search
| Gene Symbol: Idh3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031551.1 | NP_113739.1 | 12 - 13 | 1135 | 70 | |
| XM_006229551.1 | XP_006229613.1 | 10 - 11 | 1557 | 70 | ||
| XM_006229552.2 | XP_006229614.1 | 11 - 12 | 1140 | 70 | ||
| XM_006229553.2 | XP_006229615.1 | 12 - 13 | 1160 | 70 | ||
| XM_006229554.1 | XP_006229616.1 | 11 - 12 | 1087 | 70 | ||
| GenBank mRNA | BC083688.1 | - | 12 - 13 | 1135 | 7189278 | 70 |
| FQ213717.1 | - | 12 - 13 | 1014 | 70 | ||
| FQ214482.1 | - | 12 - 13 | 1102 | 70 | ||
| FQ214526.1 | - | 12 - 13 | 1101 | 70 | ||
| FQ214718.1 | - | 12 - 13 | 1152 | 70 | ||
| FQ214896.1 | - | 12 - 13 | 1085 | 70 | ||
| FQ217943.1 | - | 2 - 3 | 162 | 70 | ||
| FQ226989.1 | - | 11 - 12 | 1012 | 70 | ||
| X74125.1 | - | 12 - 13 | 1061 | 70 | ||
Set Membership: |
|