Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01759158_m1
          See other RGD1309106 GE Assays ›
          Gene Symbol:
          RGD1309106
          Gene Name:
          similar to hypothetical protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 79856479 - 79899479 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01759158_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1309106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014130.1 NP_001014152.1 6 - 7 528 74
          XM_006250168.2 XP_006250230.1 6 - 7 517 74
          XM_006250169.2 XP_006250231.1 6 - 7 517 74
          XM_006250170.2 XP_006250232.1 6 - 7 731 74
          XM_006250171.2 XP_006250233.1 6 - 7 478 74
          GenBank mRNA BC083573.1 - 6 - 7 528 7112776 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001762.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1309106

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46634 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098606 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87776 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8722 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769265 - -
          Affymetrix Rat Gene 1.0 ST Array 10769269 - -
          Affymetrix Rat Gene 1.0 ST Array 10769266 - -
          Affymetrix Rat Gene 1.0 ST Array 10769263 - -
          Affymetrix Rat Gene 1.0 ST Array 10769267 - -
          Affymetrix Rat Gene 1.0 ST Array 10769264 - -
          Affymetrix Rat Gene 1.0 ST Array 10769270 - -
          Affymetrix Rat Gene 1.0 ST Array 10769268 - -
          Affymetrix Rat Gene 1.1 ST Array 10769262 - -
          Affymetrix Rat Clariom S Assay PSR1300012373.rn.1 TC1300001762.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012374.rn.1 TC1300001762.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012376.rn.1 TC1300001762.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10769262 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21980314 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358636 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline