Search
Search
| Gene Symbol: Ric8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100520.1 | NP_001093990.1 | 8 - 9 | 1581 | 60 | |
| GenBank mRNA | AY177754.1 | - | 8 - 9 | 1338 | 60 | |
| BC160852.1 | - | 8 - 9 | 1581 | 8364542 | 60 | |
| FQ223245.1 | - | 1276 | 60 | |||
Set Membership: |
|