Search
Search

Rtn4ip1| Gene Symbol: Rtn4ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107644.3 | NP_001101114.1 | 2 - 3 | 819 | 78 | |
| XM_006256589.2 | XP_006256651.1 | 2 - 3 | 840 | 78 | ||
| XM_006256590.2 | XP_006256652.1 | 2 - 3 | 840 | 78 | ||
| XM_008772999.1 | XP_008771221.1 | 3 - 4 | 508 | 78 | ||
| GenBank mRNA | BC082054.1 | - | 2 - 3 | 840 | 78 | |
| BC166465.1 | - | 2 - 3 | 823 | 9030981 | 78 | |
Set Membership: |
|