Search
Search
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106381.1 | NP_001099851.1 | 6 - 7 | 1054 | 62 | |
| XM_006256338.2 | XP_006256400.1 | 6 - 7 | 1074 | 62 | ||
| GenBank mRNA | BC168843.1 | - | 5 - 6 | 846 | 9089070 | 62 |
Set Membership: |
|