Search
Search
| Gene Symbol: Ffar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005877.1 | NP_001005877.1 | 1 | 942 | 122 | |
| XM_006228821.2 | XP_006228883.1 | 2 | 1694 | 122 | ||
| XM_006228822.2 | XP_006228884.1 | 3 | 1288 | 122 | ||
| XM_008759133.1 | XP_008757355.1 | 3 | 1388 | 122 | ||
| XM_008759134.1 | XP_008757356.1 | 3 | 1349 | 122 | ||
| XM_008759135.1 | XP_008757357.1 | 3 | 1066 | 122 | ||
| GenBank mRNA | AB106675.1 | - | 1 | 942 | 122 | |
Set Membership: |
|