Search
Search
| Gene Symbol: Oas1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009489.1 | NP_001009489.1 | 2 - 3 | 527 | 124 | |
| XM_006249400.2 | XP_006249462.1 | 2 - 3 | 503 | 124 | ||
| XM_006249401.2 | XP_006249463.1 | 2 - 3 | 503 | 124 | ||
| GenBank mRNA | AY196696.1 | - | 2 - 3 | 527 | 124 | |
| BC088437.1 | - | 2 - 3 | 476 | 7125462 | 124 | |
Set Membership: |
|