Search
Search
| Gene Symbol: Ncald | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024371.1 | NP_001019542.1 | 3 - 4 | 650 | 87 | |
| XM_008765456.1 | XP_008763678.1 | 4 - 5 | 640 | 87 | ||
| GenBank mRNA | BC087105.1 | - | 3 - 4 | 650 | 7193019 | 87 |
| FQ214436.1 | - | 659 | 87 | |||
Set Membership: |
|