Search
Search
| Gene Symbol: Ddc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017481.1 | NP_001017481.1 | 2 | 1636 | 77 | |
| XM_006247814.2 | XP_006247876.1 | 2 | 2211 | 77 | ||
| XM_006247815.2 | XP_006247877.1 | 3 | 3688 | 77 | ||
| XM_006247816.2 | XP_006247878.1 | 3 | 3554 | 77 | ||
| XM_006247817.2 | XP_006247879.2 | 2 | 2889 | 77 | ||
| XM_008768384.1 | XP_008766606.1 | 3 | 3724 | 77 | ||
| XM_008768385.1 | XP_008766607.1 | 3 | 3953 | 77 | ||
| XM_008768386.1 | XP_008766608.1 | 3 | 3553 | 77 | ||
| XM_008768387.1 | XP_008766609.1 | 2 | 1728 | 77 | ||
| GenBank mRNA | BC079213.1 | - | 2 | 1636 | 7131836 | 77 |
Set Membership: |
|