Search
Search

LOC288521| Gene Symbol: LOC288521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013868.1 | NP_001013890.1 | 947 | 116 | ||
| GenBank mRNA | BC079301.1 | - | 947 | 7134538 | 116 | |
Set Membership: |
|