Search
Search

LOC689713| Gene Symbol: LOC689713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047978.1 | NP_001041443.1 | 2 - 3 | 118 | 62 | |
| GenBank mRNA | AY539926.1 | - | 2 - 3 | 118 | 62 | |
Set Membership: |
|