Search
Search
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270931.1 | NP_001257860.1 | 4 - 5 | 568 | 53 | |
| NM_001270933.1 | NP_001257862.1 | 4 - 5 | 568 | 53 | ||
| NM_022180.2 | NP_071516.2 | 3 - 4 | 528 | 53 | ||
| XM_006235515.2 | XP_006235577.1 | 3 - 4 | 523 | 53 | ||
| XM_006235516.2 | XP_006235578.1 | 3 - 4 | 523 | 53 | ||
| GenBank mRNA | CK483260.1 | - | 3 - 4 | 357 | 53 | |
| D10554.1 | - | 3 - 4 | 404 | 53 | ||
| EF193390.1 | - | 3 - 4 | 349 | 53 | ||
| EF193392.1 | - | 3 - 4 | 330 | 53 | ||
| FJ608816.1 | - | 4 - 5 | 568 | 53 | ||
| FJ608817.1 | - | 4 - 5 | 568 | 53 | ||
| FJ608818.1 | - | 4 - 5 | 568 | 53 | ||
| FM042560.1 | - | 3 - 4 | 533 | 53 | ||
| X57133.1 | - | 3 - 4 | 419 | 53 | ||
Set Membership: |
|