Search
Search
| Gene Symbol: Ptms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031975.2 | NP_114181.1 | 1 - 2 | 384 | 62 | |
| GenBank mRNA | BC167753.1 | - | 1 - 2 | 374 | 7371571 | 62 |
| X16481.1 | - | 1 - 2 | 167 | 62 | ||
Set Membership: |
|