Search
Search
| Gene Symbol: Phb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031851.2 | NP_114039.1 | 7 | 1701 | 69 | |
| GenBank mRNA | BC072518.1 | - | 7 | 1701 | 7105633 | 69 |
| FQ229103.1 | - | 7 | 1703 | 69 | ||
| M61219.1 | - | 6 | 1650 | 69 | ||
Set Membership: |
|