Search
Search

LOC100911196| Gene Symbol: LOC100911196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228306.2 | XP_006228368.1 | 1 | 1672 | 86 | |
| XM_008759010.1 | XP_008757232.1 | 1 | 2203 | 86 | ||
Set Membership: |
|