Search
Search
| Gene Symbol: Degs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017457.2 | NP_001017457.1 | 2 - 3 | 897 | 64 | |
| GenBank mRNA | AJ938082.1 | - | 2 - 3 | 830 | 64 | |
| BC098701.1 | - | 2 - 3 | 897 | 7387212 | 64 | |
Set Membership: |
|