Search
Search
| Gene Symbol: Mtif2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004254.1 | NP_001004254.1 | 3 - 4 | 344 | 71 | |
| GenBank mRNA | BC079421.1 | - | 3 - 4 | 344 | 7113831 | 71 |
| FQ224859.1 | - | 3 - 4 | 412 | 71 | ||
Set Membership: |
|