Search
Search
| Gene Symbol: DNAJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244163.1 | NP_001231092.1 | 2 - 3 | 264 | 75 | |
| XM_021063987.1 | XP_020919646.1 | 298 | 75 | |||
| GenBank mRNA | AK234259.1 | - | 2 - 3 | 273 | 75 | |
| AK234897.1 | - | 2 - 3 | 242 | 75 | ||
| AK351953.1 | - | 2 - 3 | 264 | 75 | ||
| AY609413.1 | - | 2 - 3 | 224 | 75 | ||
Set Membership: |
|