Search
Search

ubp1| Gene Symbol: ubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011473.1 | NP_001011473.1 | 14 - 15 | 1428 | 60 | |
| XM_012971725.2 | XP_012827179.1 | 14 - 15 | 1485 | 60 | ||
| GenBank mRNA | BC088609.1 | - | 14 - 15 | 1428 | 60 | |
Set Membership: |
|