Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aph1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015843.1 | NP_001015843.1 | 3 - 4 | 377 | 74 | |
XM_012953193.2 | XP_012808647.1 | 3 - 4 | 409 | 74 | ||
GenBank mRNA | BC090132.1 | - | 3 - 4 | 377 | 74 | |
CT030063.1 | - | 3 - 4 | 371 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |