Search
Search

dpepe| Gene Symbol: dpepe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016448.2 | NP_001016448.1 | 4 - 5 | 373 | 76 | |
| XM_012970425.2 | XP_012825879.1 | 4 - 5 | 425 | 76 | ||
| XM_012970426.2 | XP_012825880.1 | 4 - 5 | 435 | 76 | ||
| XM_012970427.2 | XP_012825881.1 | 4 - 5 | 412 | 76 | ||
| XM_018097107.1 | XP_017952596.1 | 4 - 5 | 386 | 76 | ||
| GenBank mRNA | BC167317.1 | - | 4 - 5 | 423 | 76 | |
| BC170743.1 | - | 4 - 5 | 351 | 76 | ||
| BC170745.1 | - | 4 - 5 | 351 | 76 | ||
| CR760999.2 | - | 4 - 5 | 373 | 76 | ||
Set Membership: |
|