Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016183.3 | NP_001016183.1 | 2 - 3 | 250 | 57 | |
GenBank mRNA | BC118696.1 | - | 2 - 3 | 250 | 57 | |
CR761828.2 | - | 2 - 3 | 277 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |