Search
Search

tpm4| Gene Symbol: tpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017087.3 | NP_001017087.1 | 1 - 2 | 448 | 62 | |
| XM_012963264.2 | XP_012818718.1 | 1 - 2 | 639 | 62 | ||
| XM_012963267.2 | XP_012818721.1 | 2 - 3 | 476 | 62 | ||
| XM_012963269.2 | XP_012818723.1 | 1 - 2 | 644 | 62 | ||
| GenBank mRNA | BC135407.1 | - | 1 - 2 | 448 | 62 | |
| CR761414.2 | - | 1 - 2 | 304 | 62 | ||
Set Membership: |
|