Search
Search

casz1| Gene Symbol: casz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012966965.2 | XP_012822419.1 | 3 - 4 | 1887 | 200 | |
| XM_012966967.2 | XP_012822421.1 | 3 - 4 | 1887 | 173 | ||
| XM_012966970.2 | XP_012822424.1 | 6 - 7 | 1684 | 173 | ||
| XM_018095834.1 | XP_017951323.1 | 3 - 4 | 1740 | 173 | ||
| XM_018095835.1 | XP_017951324.1 | 3 - 4 | 1890 | 200 | ||
| XM_018095836.1 | XP_017951325.1 | 3 - 4 | 1889 | 173 | ||
| GenBank mRNA | EF666979.1 | - | 3 - 4 | 1809 | 173 | |
| EF666980.1 | - | 6 - 7 | 1695 | 173 | ||
Set Membership: |
|