Search
Search

MIR922
RUBCNGACTTCACCATCAACACACATTAAC[G/A]GGGGGAGGGGGGCAGCACACTAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MIR922 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MIR922 - microRNA 922 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| RUBCN - RUN and cysteine rich domain containing beclin 1 interacting protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001145642.3 | 4527 | UTR 3 | NP_001139114.1 | |||
| NM_014687.2 | 4527 | UTR 3 | NP_055502.1 | |||
| XM_005269374.3 | 4527 | UTR 3 | XP_005269431.1 | |||
| XM_006713827.3 | 4527 | UTR 3 | XP_006713890.1 | |||
| XM_006713828.3 | 4527 | UTR 3 | XP_006713891.1 | |||
| XM_006713829.3 | 4527 | UTR 3 | XP_006713892.1 | |||
| XM_006713830.3 | 4527 | UTR 3 | XP_006713893.1 | |||
| XM_006713831.3 | 4527 | UTR 3 | XP_006713894.1 | |||
| XM_017007543.1 | 4527 | UTR 3 | XP_016863032.1 | |||
| XM_017007544.1 | 4527 | UTR 3 | XP_016863033.1 | |||
| XM_017007545.1 | 4527 | UTR 3 | XP_016863034.1 | |||