Search
Search

ACTA2
ACTA2-AS1ACATCTGCTGGAAGGTGGACAGAGA[A/G]GCCAGGATGGAGCCACCGATCCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ACTA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ACTA2 - actin, alpha 2, smooth muscle, aorta | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001141945.2 | 1529 | Silent Mutation | GCC,GCT | A,A 349 | NP_001135417.1 | |
| NM_001320855.1 | 1529 | Silent Mutation | GCC,GCT | A,A 349 | NP_001307784.1 | |
| NM_001613.2 | 1529 | Silent Mutation | NP_001604.1 | |||
| ACTA2-AS1 - ACTA2 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||