Search
Search

CDH11AGCCTCTGGCAAAGGGCCTGCCTGA[C/A]AAATAGTAAGAACTCAATAACTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CDH11 - cadherin 11 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001308392.1 | Intron | NP_001295321.1 | ||||
| NM_001797.3 | Intron | NP_001788.2 | ||||
| XM_005255761.3 | Intron | XP_005255818.1 | ||||
| XM_005255762.2 | Intron | XP_005255819.1 | ||||
| XM_005255763.2 | Intron | XP_005255820.1 | ||||
| XM_011522803.2 | Intron | XP_011521105.1 | ||||
| XM_017022830.1 | Intron | XP_016878319.1 | ||||