Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCAGCTCCTTGAGGAGAATGA[C/T]CAGCTGATCCGCTGTATTGTGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 161565 MIM: 604028 MIM: 606473 | ||||||||||||||||||||
Literature Links: |
NKTR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKTR - natural killer cell triggering receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC22C - SEC22 homolog C, vesicle trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201572.1 | 1307 | Intron | NP_001188501.1 | |||
NM_001201584.1 | 1307 | Intron | NP_001188513.1 | |||
NM_004206.3 | 1307 | Intron | NP_004197.1 | |||
NM_032970.3 | 1307 | Intron | NP_116752.1 | |||
XM_005265533.1 | 1307 | Intron | XP_005265590.1 | |||
XM_005265534.1 | 1307 | Intron | XP_005265591.1 | |||
XM_005265535.1 | 1307 | Intron | XP_005265592.1 | |||
XM_011534212.1 | 1307 | Intron | XP_011532514.1 | |||
XM_017007438.1 | 1307 | Intron | XP_016862927.1 | |||
XM_017007439.1 | 1307 | Intron | XP_016862928.1 |
SS18L2 - SS18 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016305.3 | 1307 | Intron | NP_057389.1 | |||
XM_017006548.1 | 1307 | Silent Mutation | GAC,GAT | D,D 29 | XP_016862037.1 | |
XM_017006549.1 | 1307 | Intron | XP_016862038.1 |