Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCAGTGCTGGTGAAGGTGGACT[C/T]GGAGGAGGAGGGGAACTCGCCCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603493 | ||||||||||||||||||||
Literature Links: |
KCNK5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNK5 - potassium two pore domain channel subfamily K member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003740.3 | 1769 | Missense Mutation | AAG,GAG | K,E 469 | NP_003731.1 | |
XM_005249456.1 | 1769 | Missense Mutation | AAG,GAG | K,E 466 | XP_005249513.1 | |
XM_006715235.2 | 1769 | Missense Mutation | AAG,GAG | K,E 287 | XP_006715298.1 |