Search
Search

ABCB8
ASIC3
CDK5
SLC4A2GATGCACGGGCTGGGCCACGTCTTC[A/G]GGCCAGGCAGCCTGAGCCTGCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ABCB8 - ATP binding cassette subfamily B member 8 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| ASIC3 - acid sensing ion channel subunit 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004769.3 | 694 | Missense Mutation | AGG,GGG | R,G 34 | NP_004760.1 | |
| NM_020321.3 | 694 | Missense Mutation | AGG,GGG | R,G 34 | NP_064717.1 | |
| NM_020322.3 | 694 | Missense Mutation | AGG,GGG | R,G 34 | NP_064718.1 | |
| CDK5 - cyclin dependent kinase 5 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SLC4A2 - solute carrier family 4 member 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||