Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGCGAGTGTTCGCTGTCGGGGT[C/T]TGATCCGTGGATACAGCTGGGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606455 MIM: 613504 | ||||||||||||||||||||
Literature Links: |
PPP1R13B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R13B - protein phosphatase 1 regulatory subunit 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015316.2 | 4040 | Missense Mutation | AAA,AGA | K,R 1083 | NP_056131.2 | |
XM_005267487.4 | 4040 | Missense Mutation | AAA,AGA | K,R 1151 | XP_005267544.1 | |
XM_011536593.2 | 4040 | Missense Mutation | AAA,AGA | K,R 1186 | XP_011534895.1 | |
XM_017021116.1 | 4040 | Missense Mutation | AAA,AGA | K,R 1118 | XP_016876605.1 | |
XM_017021117.1 | 4040 | Missense Mutation | AAA,AGA | K,R 1115 | XP_016876606.1 | |
XM_017021118.1 | 4040 | Missense Mutation | AAA,AGA | K,R 1115 | XP_016876607.1 | |
XM_017021119.1 | 4040 | Missense Mutation | AAA,AGA | K,R 866 | XP_016876608.1 |
ZFYVE21 - zinc finger FYVE-type containing 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |