Search
Search

CHD3
NAA38CAGCTGCTGTCTCGTCGGCCACCCC[A/C]ATAGCACCCTCCGGACCCCCCGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CHD3 - chromodomain helicase DNA binding protein 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001005271.2 | 919 | Silent Mutation | CCA,CCC | P,P 292 | NP_001005271.2 | |
| NM_001005273.2 | 919 | Silent Mutation | CCA,CCC | P,P 233 | NP_001005273.1 | |
| NM_005852.3 | 919 | Silent Mutation | CCA,CCC | P,P 233 | NP_005843.2 | |
| XM_005256427.4 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_005256484.1 | |
| XM_005256428.4 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_005256485.1 | |
| XM_005256429.4 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_005256486.1 | |
| XM_005256431.4 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_005256488.1 | |
| XM_006721423.3 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_006721486.1 | |
| XM_006721424.3 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_006721487.1 | |
| XM_006721428.3 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_006721491.1 | |
| XM_017024061.1 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_016879550.1 | |
| XM_017024062.1 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_016879551.1 | |
| XM_017024063.1 | 919 | Silent Mutation | CCA,CCC | P,P 233 | XP_016879552.1 | |
| XM_017024064.1 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_016879553.1 | |
| XM_017024065.1 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_016879554.1 | |
| XM_017024066.1 | 919 | Silent Mutation | CCA,CCC | P,P 213 | XP_016879555.1 | |
| XM_017024067.1 | 919 | Silent Mutation | CCA,CCC | P,P 222 | XP_016879556.1 | |
| XM_017024068.1 | 919 | Silent Mutation | CCA,CCC | P,P 213 | XP_016879557.1 | |
| XM_017024069.1 | 919 | Silent Mutation | CCA,CCC | P,P 65 | XP_016879558.1 | |
| XM_017024070.1 | 919 | Silent Mutation | CCA,CCC | P,P 292 | XP_016879559.1 | |
| NAA38 - N(alpha)-acetyltransferase 38, NatC auxiliary subunit | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||