Search
Search

THEGGGCAAGCTCATAGAGGCGAGGGGAC[A/G]CCTGTCGTATCCCTCGTTGAGGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
THEG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| THEG - theg spermatid protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_016585.4 | 1808 | Missense Mutation | NP_057669.1 | |||
| NM_199202.2 | 1808 | Missense Mutation | NP_954672.1 | |||
| XM_011528049.2 | 1808 | Missense Mutation | XP_011526351.1 | |||
| XM_011528050.2 | 1808 | Missense Mutation | XP_011526352.1 | |||
| XM_011528051.2 | 1808 | Missense Mutation | XP_011526353.1 | |||
| XM_011528052.2 | 1808 | Intron | XP_011526354.1 | |||