Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACCGCATCCTGTGCTCATACA[A/C]CACCAACATTGAAGAGCTCTTTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605522 MIM: 610241 | ||||||||||||||||||||
Literature Links: |
LMBR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMBR1 - limb development membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022458.3 | 962 | Intron | NP_071903.2 | |||
XM_005249555.2 | 962 | Intron | XP_005249612.1 | |||
XM_005249558.2 | 962 | Intron | XP_005249615.1 | |||
XM_017012507.1 | 962 | Intron | XP_016867996.1 | |||
XM_017012508.1 | 962 | Intron | XP_016867997.1 | |||
XM_017012509.1 | 962 | Intron | XP_016867998.1 | |||
XM_017012510.1 | 962 | Intron | XP_016867999.1 | |||
XM_017012511.1 | 962 | Intron | XP_016868000.1 | |||
XM_017012512.1 | 962 | Intron | XP_016868001.1 | |||
XM_017012513.1 | 962 | Intron | XP_016868002.1 | |||
XM_017012514.1 | 962 | Intron | XP_016868003.1 | |||
XM_017012515.1 | 962 | Intron | XP_016868004.1 | |||
XM_017012516.1 | 962 | Intron | XP_016868005.1 | |||
XM_017012517.1 | 962 | Intron | XP_016868006.1 | |||
XM_017012518.1 | 962 | Intron | XP_016868007.1 |
RNF32 - ring finger protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184996.1 | 962 | Missense Mutation | AAC,ACC | N,T 241 | NP_001171925.1 | |
NM_001184997.1 | 962 | Missense Mutation | AAC,ACC | N,T 241 | NP_001171926.1 | |
NM_001308273.1 | 962 | Missense Mutation | AAC,ACC | N,T 241 | NP_001295202.1 | |
NM_001308274.1 | 962 | Intron | NP_001295203.1 | |||
NM_030936.3 | 962 | Missense Mutation | AAC,ACC | N,T 241 | NP_112198.1 | |
XM_005249522.4 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_005249579.1 | |
XM_011515804.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514106.1 | |
XM_011515805.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514107.1 | |
XM_011515806.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514108.1 | |
XM_011515807.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514109.1 | |
XM_011515808.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514110.1 | |
XM_011515809.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514111.1 | |
XM_011515810.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514112.1 | |
XM_011515811.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514113.1 | |
XM_011515812.2 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_011514114.1 | |
XM_011515813.2 | 962 | Missense Mutation | AAC,ACC | N,T 228 | XP_011514115.1 | |
XM_017011753.1 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_016867242.1 | |
XM_017011754.1 | 962 | Missense Mutation | AAC,ACC | N,T 241 | XP_016867243.1 | |
XM_017011755.1 | 962 | Intron | XP_016867244.1 |