Search
Search

CDK1ATGGTTGGTTAGTTCTAGATCACTA[A/G]TCTGTTATTTGACTTATTCACGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
CDK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| CDK1 - cyclin dependent kinase 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001170406.1 | Intron | NP_001163877.1 | ||||
| NM_001170407.1 | Intron | NP_001163878.1 | ||||
| NM_001320918.1 | Intron | NP_001307847.1 | ||||
| NM_001786.4 | Intron | NP_001777.1 | ||||
| NM_033379.4 | Intron | NP_203698.1 | ||||
| XM_005270303.3 | Intron | XP_005270360.1 | ||||