Search
Search

BCL10
C1orf52
LOC646626TTCAGAGAAATTACTCTCATCTGAA[T/-]TTGATCTGGAGAGGTTGTTCGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
BCL10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| BCL10 - B-cell CLL/lymphoma 10 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001320715.1 | 831 | Frame Shift Delete | AAT,ATT | N,I 126 | NP_001307644.1 | |
| NM_003921.4 | 831 | Frame Shift Delete | AAT,ATT | N,I 137 | NP_003912.1 | |
| XM_011542397.2 | 831 | Frame Shift Delete | AAT,ATT | N,I 190 | XP_011540699.1 | |
| XM_011542398.2 | 831 | Frame Shift Delete | AAT,ATT | N,I 179 | XP_011540700.1 | |
| XM_011542399.2 | 831 | Frame Shift Delete | AAT,ATT | N,I 119 | XP_011540701.1 | |
| C1orf52 - chromosome 1 open reading frame 52 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC646626 - uncharacterized LOC646626 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||