Search
Search

DDR2TTAGGGTAACAGTGGGATAATTCTT[T/C]TCTGTATGTAGTTATGTCACCAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
DDR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| DDR2 - discoidin domain receptor tyrosine kinase 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001014796.1 | Intron | NP_001014796.1 | ||||
| NM_006182.2 | Intron | NP_006173.2 | ||||
| XM_006711344.3 | Intron | XP_006711407.1 | ||||
| XM_011509586.2 | Intron | XP_011507888.1 | ||||
| XM_011509587.2 | Intron | XP_011507889.1 | ||||
| XM_011509588.2 | Intron | XP_011507890.1 | ||||