Search
Search

CASP9
DNAJC16AAAAATTTCTGAATTTGTTGCTAAC[A/G]TTTTAAGCCAGGGAATTTGACAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
CASP9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| CASP9 - caspase 9 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001229.4 | Intron | NP_001220.2 | ||||
| NM_001278054.1 | Intron | NP_001264983.1 | ||||
| NM_032996.3 | Intron | NP_127463.2 | ||||
| XM_005246014.2 | Intron | XP_005246071.1 | ||||
| XM_011542273.2 | Intron | XP_011540575.1 | ||||
| DNAJC16 - DnaJ heat shock protein family (Hsp40) member C16 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||