Search
Search

IL1F10
IL36RNTCTGCCACCCACCGCCCCCAATCTA[T/A]CTTGGCTCACTCGCTCTGGGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1F10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| IL1F10 - interleukin 1 family member 10 (theta) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| IL36RN - interleukin 36 receptor antagonist | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_012275.2 | 2450 | UTR 3 | NP_036407.1 | |||
| NM_173170.1 | 2450 | UTR 3 | NP_775262.1 | |||