Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609793 | ||||||||||||||||||||
Literature Links: |
LINGO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINGO2 - leucine rich repeat and Ig domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258282.1 | 17380 | Intron | NP_001245211.1 | |||
NM_152570.2 | 17380 | Intron | NP_689783.1 | |||
XM_011517719.1 | 17380 | UTR 3 | XP_011516021.1 | |||
XM_011517724.2 | 17380 | UTR 3 | XP_011516026.1 | |||
XM_011517728.2 | 17380 | UTR 3 | XP_011516030.1 | |||
XM_017014303.1 | 17380 | UTR 3 | XP_016869792.1 | |||
XM_017014304.1 | 17380 | UTR 3 | XP_016869793.1 | |||
XM_017014305.1 | 17380 | UTR 3 | XP_016869794.1 | |||
XM_017014306.1 | 17380 | UTR 3 | XP_016869795.1 | |||
XM_017014307.1 | 17380 | UTR 3 | XP_016869796.1 |