Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECT2L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| ECT2L - epithelial cell transforming 2 like | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001077706.2 | Intron | NP_001071174.1 | ||||
| NM_001195037.2 | Intron | NP_001181966.1 | ||||
| XM_006715472.3 | Intron | XP_006715535.1 | ||||
| XM_011535795.2 | Intron | XP_011534097.1 | ||||
| XM_011535797.2 | Intron | XP_011534099.1 | ||||
| XM_017010828.1 | Intron | XP_016866317.1 | ||||
| XM_017010829.1 | Intron | XP_016866318.1 | ||||
| XM_017010830.1 | Intron | XP_016866319.1 | ||||