Search
Search

CDK7AATACTATATCTTATTTTTATTTTG[G/T]TTATTTTTTTTGAGACAGAGTCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
CDK7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| CDK7 - cyclin dependent kinase 7 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001324069.1 | Intron | NP_001310998.1 | ||||
| NM_001324070.1 | Intron | NP_001310999.1 | ||||
| NM_001324071.1 | Intron | NP_001311000.1 | ||||
| NM_001324072.1 | Intron | NP_001311001.1 | ||||
| NM_001324074.1 | Intron | NP_001311003.1 | ||||
| NM_001324075.1 | Intron | NP_001311004.1 | ||||
| NM_001324077.1 | Intron | NP_001311006.1 | ||||
| NM_001324078.1 | Intron | NP_001311007.1 | ||||
| NM_001799.3 | Intron | NP_001790.1 | ||||
| XM_011543094.2 | Intron | XP_011541396.1 | ||||